• USA Home
  • Search Results

  • tetraethylenepentamine1893011257211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetraethylene pentamine

2 Products
Synonym:   1,4,7,10,13-Pentaazatridecane, Tetraethylene pentamine
Image of Tetraethylene pentamine
CAS Number:
Empirical Formula (Hill Notation):
 C8H23N5
Molecular Weight:
 189.30
 EC Index Number:
 203-986-2
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Potency
  Special Grade
  Vapor pressure
Show More