• USA Home
  • Search Results

  • tetrafluoroboricaciddiethylethercomplex161936796982811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetrafluoroboric acid diethyl ether complex

1 Products
Synonym:   Fluoroboric acid diethyl ether complex
Image of Tetrafluoroboric acid diethyl ether complex
CAS Number:
Linear Formula:
 HBF4 · O(CH2CH3)2
Molecular Weight:
 161.93
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More