• USA Home
  • Search Results

  • tetrahydrofolicacid4454313516011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetrahydrofolic acid

1 Products
Synonym:   5,6,7,8-Tetrahydropteroyl-L-glutamic acid, THF
Image of Tetrahydrofolic acid
CAS Number:
Empirical Formula (Hill Notation):
 C19H23N7O6
Molecular Weight:
 445.43
 Beilstein/REAXYS Number:
 9238187
 EC Number:
 MDL number:
 NACRES:
 NA.51
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More