• USA Home
  • Search Results

  • tetrahydrofuran721110999911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetrahydrofuran

23 Products
Synonym:   Butylene oxide, Oxolane, THF, Tetramethylene oxide
Image of Tetrahydrofuran
CAS Number:
Empirical Formula (Hill Notation):
 C4H8O
Molecular Weight:
 72.11
 Beilstein/REAXYS Number:
 102391
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Package quantity
  Density
  Restriction
  Suitability
  Quality clarifier
  Refractive Index
  Application(s)
  UV-Absorption Condition
  Purity Grade
  pH Value
  Transition temperature
  Color
  Solubility
  Absorption
  Additive
  UV-Absorption Values
  Assay
Show More

Product #
Description
401757 Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free,
34865 Tetrahydrofuran, inhibitor-free, for HPLC, ≥99.9%,
186562 Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%,
439215 Tetrahydrofuran, for HPLC, ≥99.9%, inhibitor-free,
360589 Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%,
178810 Tetrahydrofuran, ReagentPlus®, ≥99.0%, contains 250 ppm BHT as inhibitor,
87368 Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, Reag. Ph. Eur., ≥99.9%,
676764 Tetrahydrofuran, ACS reagent, ≥99.0%, contains 250 ppm BHT as inhibitor,
78445 Tetrahydrofuran, analytical standard,
87369 Tetrahydrofuran, Selectophore, ≥99.5%,
437638 Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%,
1.08101 Tetrahydrofuran, for liquid chromatography LiChrosolv®,
1.09731 Tetrahydrofuran, for analysis EMSURE® ACS,Reag. Ph Eur,
1.08114 Tetrahydrofuran, EMPLURA®,
1.08110 Tetrahydrofuran, for spectroscopy Uvasol®,
1.07025 Tetrahydrofuran, for analysis EMPARTA® ACS,
1.08107 Tetrahydrofuran, for DNA and peptide synthesis (max. 0.005% H₂O),
TX0280 Tetrahydrofuran, Meets ACS Specifications GR ACS,
PHR1561 Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material,
900636 Tetrahydrofuran, anhydrous, inhibitor-free, ZerO2®, ≥99.9%,
RTC000088 Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material,
TX0277 Tetrahydrofuran, anhydrous, septum sealed-bottle DriSolv,
TX0284 Tetrahydrofuran, anhydrous, septum sealed-bottle DriSolv,