• USA Home
  • Search Results

  • tetrahydrofurfurylamine10115479529311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetrahydrofurfurylamine

1 Products
Synonym:   2-(Aminomethyl)tetrahydrofuran
Image of Tetrahydrofurfurylamine
CAS Number:
Empirical Formula (Hill Notation):
 C5H11NO
Molecular Weight:
 101.15
 Beilstein/REAXYS Number:
 102729
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More