• USA Home
  • Search Results

  • tetrahydrofurfurylchloride12058300384711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetrahydrofurfuryl chloride

1 Products
Synonym:   2-(Chloromethyl)tetrahydrofuran
Image of Tetrahydrofurfuryl chloride
CAS Number:
Empirical Formula (Hill Notation):
 C5H9ClO
Molecular Weight:
 120.58
 Beilstein/REAXYS Number:
 102716
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More