• USA Home
  • Search Results

  • tetrakisdecylammoniumbromide659011493742911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetrakis(decyl)ammonium bromide

2 Products
Synonym:   TDAB, Tetra-decylammonium bromide
Image of Tetrakis(decyl)<WBR>ammonium bromide
CAS Number:
Linear Formula:
 [CH3(CH2)9]4N(Br)
Molecular Weight:
 659.01
 Beilstein/REAXYS Number:
 5842649
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Molecular weight
  ID clarifier
  UV-Absorption Values
Show More