• USA Home
  • Search Results

  • tetrakistriphenylphosphinenickel01107841513382111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetrakis(triphenylphosphine)nickel(0)

1 Products
Synonym:   Ni(PPh3)4, Nickel-tetrakis(triphenylphosphine)
Image of Tetrakis(triphenylphosphine)<WBR>nickel(0)
CAS Number:
Linear Formula:
 [(C6H5)3P]4Ni
Molecular Weight:
 1107.84
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More