• USA Home
  • Search Results

  • tetramethylsilane88227576311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetramethylsilane

5 Products
Synonym:   TMS, TMS
Image of Tetramethylsilane
CAS Number:
Linear Formula:
 (CH3)4Si
Molecular Weight:
 88.22
 EC Index Number
 200-899-1
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Refractive Index
  Assay
Show More