• USA Home
  • Search Results

  • tetramisolehydrochloride240751659580511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(−)-Tetramisole hydrochloride

1 Products
Synonym:   (S)-(−)-6-Phenyl-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole hydrochloride, L(−)-2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole hydrochloride, Levamisole hydrochloride
Image of (−)-Tetramisole hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C11H12N2S · HCl
Molecular Weight:
 240.75
 Beilstein/REAXYS Number:
 4358988
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More