• USA Home
  • Search Results

  • tetrasodiumpyrophosphatedecahydrate446061347236111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetrasodium Pyrophosphate Decahydrate

1 Products
Synonym:   Sodium pyrophosphate decahydrate, Sodium pyrophosphate tetrabasic decahydrate, Tetrasodium pyrophosphate
Image of Tetrasodium Pyrophosphate Decahydrate
CAS Number:
Linear Formula:
 Na4P2O7 · 10H2O
Molecular Weight:
 446.06
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More