• USA Home
  • Search Results

  • tetratetracontane61919709822811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetratetracontane

2 Products
Image of Tetratetracontane
CAS Number:
Linear Formula:
 CH3(CH2)42CH3
Molecular Weight:
 619.19
 Beilstein/REAXYS Number:
 1804576
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Anal. Standard Form
  Physical Form
  Special Grade
  Melting Point
  Assay
Show More