• USA Home
  • Search Results

  • tetrazolesolution700528894811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tetrazole solution

1 Products
Image of Tetrazole solution
CAS Number:
Empirical Formula (Hill Notation):
 CH2N4
Molecular Weight:
 70.05
 Beilstein/REAXYS Number:
 105799
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Melting Point
  Concentration
Show More