• USA Home
  • Search Results

  • thalliumibromide28429778940411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Thallium(I) bromide

2 Products
Synonym:   Thallium monobromide, Thallous bromide
Image of Thallium(I) bromide
CAS Number:
Linear Formula:
 TlBr
Molecular Weight:
 284.29
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
Show More