• USA Home
  • Search Results

  • theaflavin33digallate868703046235211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Theaflavin 3,3′-digallate

2 Products
Image of Theaflavin 3,3′-digallate
CAS Number:
Empirical Formula (Hill Notation):
 C43H32O20
Molecular Weight:
 868.70
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More