• USA Home
  • Search Results

  • theaflavin56449467005711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Theaflavin

2 Products
Synonym:   3,4,5-Trihydroxy-1,8-bis[(2R,3R)-3,5,7-trihydroxy-2-chromanyl]-6-benzo[7]annulenone, 3,4,6-Trihydroxy-1,8-bis(3α,5,7-trihydroxy-2α-chromanyl)-5H-benzocyclohepten-5-one
Image of Theaflavin
CAS Number:
Empirical Formula (Hill Notation):
 C29H24O12
Molecular Weight:
 564.49
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Application(s)
Show More