• USA Home
  • Search Results

  • theaspirane194313643172811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-Theaspirane

2 Products
Synonym:   2,6,10,10-Tetramethyl-1-oxaspiro[4.5]dec-6-ene
Image of (±)-Theaspirane
CAS Number:
Empirical Formula (Hill Notation):
 C13H22O
Molecular Weight:
 194.31
 Beilstein/REAXYS Number:
 1424383
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More