• USA Home
  • Search Results

  • thebaine3113711537711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Thebaine

1 Products
Synonym:   Paramorphine
Image of Thebaine
CAS Number:
Empirical Formula (Hill Notation):
 C19H21NO3
Molecular Weight:
 311.37
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Anal. Standard Form
  Physical Form
  Manufacturer designation
Show More

Product #
Description
T2019 Thebaine, powder,