• USA Home
  • Search Results

  • theobromine180168367011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Theobromine

5 Products
Synonym:   2,6-Dihydroxy-3,7-dimethylpurine, 3,7-Dihydro-3,7-dimethyl-1H-purine-2,6-dione, 3,7-Dimethylxanthine
Image of Theobromine
CAS Number:
Empirical Formula (Hill Notation):
 C7H8N4O2
Molecular Weight:
 180.16
 Beilstein/REAXYS Number:
 16464
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
  Application(s)
  Manufacturer designation
  Solubility
  Assay
Show More