• USA Home
  • Search Results

  • thiabendazole2012514879811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Thiabendazole

6 Products
Synonym:   2-(4-Thiazolyl)benzimidazole
Image of Thiabendazole
CAS Number:
Empirical Formula (Hill Notation):
 C10H7N3S
Molecular Weight:
 201.25
 Beilstein/REAXYS Number:
 611403
 EC Number:
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Mode of Action
  Anal. Standard Form
  Shelf Life
  Product Line
  Color
  Assay
  Activity Spectrum
  Physical Form
  Special Grade
Show More