• USA Home
  • Search Results

  • thiamphenicol356221531845311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Thiamphenicol

3 Products
Synonym:   D-threo-2,2-Dichloro-N-(β-hydroxy-α-[hydroxymethyl]-4-[methylsulfonyl]phenethyl)acetamide
Image of Thiamphenicol
CAS Number:
Empirical Formula (Hill Notation):
 C12H15Cl2NO5S
Molecular Weight:
 356.22
 Beilstein/REAXYS Number:
 2819542
 EC Number:
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Anal. Standard Form
  Activity Spectrum
  Special Grade
  Application(s)
  Solubility
Show More