• USA Home
  • Search Results

  • thidiazuron220255170755211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Thidiazuron

2 Products
Synonym:   TDZ
Image of Thidiazuron
CAS Number:
Empirical Formula (Hill Notation):
 C9H8N4OS
Molecular Weight:
 220.25
 Beilstein/REAXYS Number:
 1078092
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Product Line
  Titertest
Show More