• USA Home
  • Search Results

  • thimerosal404815464811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Thimerosal

9 Products
Synonym:   2-(Ethylmercuriomercapto)benzoic acid sodium salt, Ethylmercurithiosalicylic acid sodium salt, Mercury-([o-carboxyphenyl]thio)ethyl sodium salt, Sodium ethylmercurithiosalicylate
Image of Thimerosal
CAS Number:
Linear Formula:
 2-(C2H5HgS)C6H4CO2Na
Molecular Weight:
 404.81
 Beilstein/REAXYS Number:
 8169555
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Agency/Method
  Application(s)
  Anal. Standard Form
  Mode of Action
  Physical Form
  Special Grade
  Assay
Show More