• USA Home
  • Search Results

  • thiocticacid20633107728711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Thioctic acid

1 Products
Synonym:   (±)-α-Lipoic acid, (±)-1,2-Dithiolane-3-pentanoic acid, 6,8-Dithiooctanoic acid, DL-α-Lipoic acid, DL-6,8-Thioctic acid, Lip(S2)
Image of Thioctic acid
CAS Number:
Empirical Formula (Hill Notation):
 C8H14O2S2
Molecular Weight:
 206.33
 Beilstein/REAXYS Number:
 81853
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More