• USA Home
  • Search Results

  • thioglycolicacid92126811111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Thioglycolic acid

4 Products
Synonym:   Mercaptoacetic acid, 2-Mercaptoethanoic acid, Thioglycolic acid
Image of Thioglycolic acid
CAS Number:
Empirical Formula (Hill Notation):
 C2H4O2S
Molecular Weight:
 92.12
 EC Index Number
 200-677-4
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  pH Value
  Vapor pressure
  Transition temperature
  Potency
  Melting Point
  Density
  Special Grade
Show More