• USA Home
  • Search Results

  • thionylchloride11897771909711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Thionyl chloride

4 Products
Synonym:   Sulfurous acid dichloride, Sulfurous oxychloride
Image of Thionyl chloride
CAS Number:
Empirical Formula (Hill Notation):
 Cl2OS
Molecular Weight:
 118.97
 EC Index Number
 016-015-00-0
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Special Grade
Show More