• USA Home
  • Search Results

  • thiourea76126256611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Thiourea

8 Products
Synonym:   Sulfourea, Thiocarbamide
Image of Thiourea
CAS Number:
Linear Formula:
 NH2CSNH2
Molecular Weight:
 76.12
 Beilstein/REAXYS Number:
 605327
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Potency
  Application(s)
  Product Line
  Loss
  pH Value
  Density
  Bulk Density
  Autoignition temp
  Solubility
  Melting Point
  Assay
  Special Grade
Show More