• USA Home
  • Search Results

  • thiram2404313726811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Thiram

2 Products
Synonym:   Bis(dimethylthiocarbamoyl) disulfide, Bis(dimethylthiocarbamyl) disulfide, Tetramethylthiuram disulfide, Thiram
Image of Thiram
CAS Number:
Linear Formula:
 (CH3)2NCSS2CSN(CH3)2
Molecular Weight:
 240.43
 Beilstein/REAXYS Number:
 1725821
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Shelf Life
  Special Grade
Show More