• USA Home
  • Search Results

  • thymidine242235089511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Thymidine

3 Products
Synonym:   1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 2′-Deoxythymidine, Thymine deoxyriboside, dT
Image of Thymidine
CAS Number:
Empirical Formula (Hill Notation):
 C10H14N2O5
Molecular Weight:
 242.23
 Beilstein/REAXYS Number:
 89285
 EC Number:
 MDL number:
 NACRES:
 NA.75
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Product Line
  Biological Source
  Physical Form
  Solubility
Show More