• USA Home
  • Search Results

  • thymine126116571411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Thymine

2 Products
Synonym:   2,4-Dihydroxy-5-methylpyrimidine, 5-Methyluracil
Image of Thymine
CAS Number:
Empirical Formula (Hill Notation):
 C5H6N2O2
Molecular Weight:
 126.11
 Beilstein/REAXYS Number:
 117880
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More