• USA Home
  • Search Results

  • thymol150228983811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Thymol

10 Products
Synonym:   2-Isopropyl-5-methylphenol, 5-Methyl-2-(1-methylethyl)phenol, 5-Methyl-2-isopropylphenol, IPMP
Image of Thymol
CAS Number:
Linear Formula:
 2-[(CH3)2CH]C6H3-5-(CH3)OH
Molecular Weight:
 150.22
 Beilstein/REAXYS Number:
 1907135
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  Titertest
  Melting Point
  Anal. Standard Form
  Shelf Life
  Special Grade
  Physical Form
  Assay
Show More