• USA Home
  • Search Results

  • thymoquinone1642049091511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Thymoquinone

2 Products
Synonym:   2-Isopropyl-5-methyl-1,4-benzoquinone
Image of Thymoquinone
CAS Number:
Empirical Formula (Hill Notation):
 C10H12O2
Molecular Weight:
 164.20
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Assay
Show More