• USA Home
  • Search Results

  • thyroglobulinhuman12345901034811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Thyroglobulin human

1 Products
Synonym:   Thyroglobulin human
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.56
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
T6830 Thyroglobulin human,