• USA Home
  • Search Results

  • tiamulinfumarate609815529796611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tiamulin fumarate

3 Products
Synonym:   Thiamutilin fumarate
Image of Tiamulin fumarate
CAS Number:
Empirical Formula (Hill Notation):
 C32H51NO8S
Molecular Weight:
 609.81
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
Show More