• USA Home
  • Search Results

  • tibolone31245563053511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tibolone

2 Products
Synonym:   (7α,17α)-17-Hydroxy-7-methyl-19-norpregn-5(10)-en-20-yn-3-one, 17-Hydroxy-7α-methyl-19-nor-17α-pregn-5(10)-en-20-yn-3-one, 5(10)-Estren-7α-methyl-17α- ethynyl-17β- ol-3-one
CAS Number:
Empirical Formula (Hill Notation):
 C21H28O2
Molecular Weight:
 312.45
 EC Number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More