• USA Home
  • Search Results

  • ticarcillindisodiumsalt42839469714711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ticarcillin disodium salt

1 Products
Synonym:   (2S,5R,6R)-6-[[(2R)-Carboxy-3-thienylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid disodium salt
Image of Ticarcillin disodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C15H14N2Na2O6S2
Molecular Weight:
 428.39
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
Show More

Product #
Description
T5639 Ticarcillin disodium salt,