• USA Home
  • Search Results

  • tiliroside594522031662511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tiliroside

2 Products
Synonym:   Astragalin 6"-trans-p-coumarate, Kaempferol 3-O-β-D-(6″-O-(E)-p-coumaroyl)glucopyranoside, Potengriffioside A
Image of Tiliroside
CAS Number:
Empirical Formula (Hill Notation):
 C30H26O13
Molecular Weight:
 594.52
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Application(s)
Show More