• USA Home
  • Search Results

  • timololmaleatesalt432492692117511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Timolol maleate salt

1 Products
Synonym:   S-(−)-1-(t-Butylamino)-3-[(4-morpholino-1,2,5-thiadiazol-3-yl)oxy]-2-propanol maleate salt, Timolol hydrogen maleate
Image of Timolol maleate salt
CAS Number:
Empirical Formula (Hill Notation):
 C13H24N4O3S · C4H4O4
Molecular Weight:
 432.49
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More