• USA Home
  • Search Results

  • tiniisulfate21477748855311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tin(II) sulfate

2 Products
Synonym:   Stannous sulfate
Image of Tin(II) sulfate
CAS Number:
Linear Formula:
 SnSO4
Molecular Weight:
 214.77
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More