• USA Home
  • Search Results

  • tioconazole387716589973211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tioconazole

3 Products
Synonym:   1-[2-[(2-Chloro-3-thienyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole, Thioconazole
Image of Tioconazole
CAS Number:
Empirical Formula (Hill Notation):
 C16H13Cl3N2OS
Molecular Weight:
 387.71
 EC Number:
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Activity Spectrum
  Special Grade
  Manufacturer designation
Show More