• USA Home
  • Search Results

  • tiotropiumbromidemonohydrate4904341120731311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tiotropium bromide monohydrate

3 Products
Synonym:   (1α, 2β, 4 β, 5α, 7β)-7-[(Hydroxydi-2-thienylacetyl)oxy]-9,9-dimethyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane bromide monohydrate, 3-Oxa-9-azoniatricyclo[3.3.1.02,4]?nonane, 7-[(2-hydroxy-2,2-di-2-thienylacetyl)oxy]-9,9-dimethyl-, bromide, hydrate (1:1:1), (1α,2β,4β,5α,7β), 6β,7β-epoxy-3β-hydroxy-8-methyl-1αH,5αH-tropanium bromide monohydrate, di-2-thienylglycolate
Image of Tiotropium bromide monohydrate
CAS Number:
Empirical Formula (Hill Notation):
 C19H22BrNO4S2 · H2O
Molecular Weight:
 490.43
 MDL number:
 NACRES
 NA.77
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
Show More