• USA Home
  • Search Results

  • titaniumdioxide79871346367711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Titanium dioxide

3 Products
Synonym:   Titania, Titanium dioxide, Titanium(IV) oxide
Image of Titanium dioxide
CAS Number:
Linear Formula:
 TiO2
Molecular Weight:
 79.87
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Package quantity
Show More