• USA Home
  • Search Results

  • titaniumivoxide79871346367711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Titanium(IV) oxide

9 Products
Synonym:   Titania, Titanium dioxide
Image of Titanium(IV) oxide
CAS Number:
Linear Formula:
 TiO2
Molecular Weight:
 79.87
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Chemical Composition
  pH Value
  Surface Dimension
  Potency
  Particle Dimension
  Bulk Density
  Melting Point
  Loss
  Composite Dimension
  Agency/Method
  Physical Form
  Special Grade
Show More