• USA Home
  • Search Results

  • titaniumivoxideanatase7987131770011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Titanium(IV) oxide, anatase

3 Products
Synonym:   Titanium dioxide
Image of Titanium(IV) oxide, anatase
CAS Number:
Linear Formula:
 TiO2
Molecular Weight:
 79.87
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Particle Dimension
  Physical Form
Show More