• USA Home
  • Search Results

  • titaniumivoxysulfatesulfuricacidhydrate1234512333400911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Titanium(IV) oxysulfate - sulfuric acid hydrate

2 Products
Image of Titanium(IV) oxysulfate - sulfuric acid hydrate
CAS Number:
Linear Formula:
 TiOSO4 · xH2SO4 · yH2O
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More