• USA Home
  • Search Results

  • titaniumtetrachloride18968755045011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Titanium tetrachloride

1 Products
Synonym:   TTC, Titanium tetrachloride, Titanium(IV) chloride
Image of Titanium tetrachloride
CAS Number:
Linear Formula:
 TiCl4
Molecular Weight:
 189.68
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More