• USA Home
  • Search Results

  • titriplexiii37224638192611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Titriplex® III

2 Products
Synonym:   Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetraacetic acid disodium salt, Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Sequestrene Na2
Image of Titriplex<SUP>®</SUP> III
CAS Number:
Empirical Formula (Hill Notation):
 C10H14N2Na2O8 · 2H2O
Molecular Weight:
 372.24
 Beilstein/REAXYS Number:
 3900609
 EC Index Number
 205-358-3
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  pH Value
  Potency
  Bulk Density
  Agency/Method
Show More