• USA Home
  • Search Results

  • tolnaftate30741239896111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tolnaftate

4 Products
Synonym:   Methyl-(3-methylphenyl)carbamothioic acid O-2-naphthyl ester
Image of Tolnaftate
CAS Number:
Empirical Formula (Hill Notation):
 C19H17NOS
Molecular Weight:
 307.41
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Anal. Standard Form
  Special Grade
Show More