• USA Home
  • Search Results

  • toluene921410888311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Toluene

25 Products
Image of Toluene
CAS Number:
Linear Formula:
 C6H5CH3
Molecular Weight:
 92.14
 Beilstein/REAXYS Number:
 635760
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 39011102
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  Shelf Life
  Refractive Index
  UV-Absorption Condition
  Purity Grade
  Ice Shipping
  Restriction
  Product Line
  Agency/Method
  Viscosity
  Transition temperature
  Solubility
  Color
  Application(s)
  Special Grade
  UV-Absorption Values
  Physical Form
Show More

Product #
Description
244511 Toluene, anhydrous, 99.8%,
179418 Toluene, ACS reagent, ≥99.5%,
32249 Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC),
34866 Toluene, suitable for HPLC, 99.9%,
650579 Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%,
89680 Toluene, analytical standard,
PHR1311 Toluene, Pharmaceutical Secondary Standard; Certified Reference Material,
1.08325 Toluene, for analysis EMSURE® ACS,ISO,Reag. Ph Eur,
179965 Toluene, Laboratory Reagent, ≥99.3%,
320552 Toluene, ACS reagent, ≥99.5%,
1.00849 Toluene, for gas chromatography MS SupraSolv®,
1.08389 Toluene, for gas chromatography ECD and FID SupraSolv®,
1.08327 Toluene, for liquid chromatography LiChrosolv®,
1.08323 Toluene, EMPLURA®,
RTC000096 Toluene,
TX0737 Toluene, OmniSolv®,
1.08331 Toluene, for spectroscopy Uvasol®,
648566 Toluene, suitable for HPLC, ≥99.9%,
TX0735 Toluene, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
900522 Toluene, anhydrous, ZerO2®, 99.8%,
1.07019 Toluene, for analysis EMPARTA® ACS,
TX0750 Toluene, histology grade, practical,
1.09768 Toluene, reference substance for gas chromatography,
676756 Toluene, ACS reagent, ≥99.5%,
270377 Toluene, HPLC grade, 99.8%,