• USA Home
  • Search Results

  • toluened810019203726511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Toluene-d8

7 Products
Image of Toluene-d<SUB>8</SUB>
CAS Number:
Linear Formula:
 C6D5CD3
Molecular Weight:
 100.19
 Beilstein/REAXYS Number:
 1909300
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Explosion limit
  Special Grade
  Titertest
  Additive
  Package quantity
  Assay
  Isotopic Purity
  Melting Point
Show More